Tagboard
Tagboard, 23×30.5 cm (9×12")
Catalog # 470149-944
Supplier: DIXON TICONDEROGA COMPANY
Supplier #: P0075010
Jump to:
Product Details & Documents
Light color cardstock tagboard.
Specifications
- Description:Tagboard
- Dimensions:23×30.5 cm (9×12")