Synedra Slide
Synedra, Whole mount
470181-000
: Avantor
Genus of Diatom
- Identifying features clearly distinguishable
- Rigorous quality control standards
Needle-shaped, found in fresh water and sea water. Causes odor issues in drinking water.
- Description:Synedra
- Orientation:Whole mount