Suspension Clamp
Suspension Clamp
470105-132
: CLHK01
Easy to use hook for your support stand to hold small loads.
- Aluminum bosshead
- Rod is 10 cm long
Use to suspend a thermometer over a hot plate or for suspending specific heat specimens. Clamp holds items at a distance of 4 1/2" from the support rod. Aluminum hook swivels and locks at any angle.
- Description:Suspension Clamp