Orders LinkContactUsLinkComponent
Surgical Scissors
Surgical Scissors
Surgical Scissors
Surgical Scissors
Each feature a sharp point for penetration and a blunt point for situations where not desired.
Available in four sizes for most dissection needs.
 : These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
  • Description
    Length
    Material
    Blade Type
  • Surgical Scissors
    110 mm (4.5")
    Stainless steel
    Sharp/Blunt
    Each (1UN)
    Each (1UN) - $45.82
    $45.82
    Each (1UN)
       $45.82
     : 140988
  • Surgical Scissors
    110 mm (4.5")
    Stainless steel
    Sharp/Blunt
    Each (1UN)
    Each (1UN) - $53.32
    $53.32
    Each (1UN)
       $53.32
     : WLS26975-A
  • Surgical Scissors
    140 mm (5.5")
    Stainless steel
    Sharp/Blunt
    Each (1UN)
    Each (1UN) - $61.30
    $61.30
    Each (1UN)
       $61.30
     : 140940
  • Surgical Scissors
    165 mm (6.5")
    Stainless steel
    Sharp/Blunt
    Each (1UN)
    Each (1UN) - $75.69
    $75.69
    Caja de 10 (1UN)
    Caja de 10 (1UN) - $82.50
    $82.50
    Each (1UN)
       $75.69
    Caja de 10 (1UN)
       $82.50
     : 140990
  • Surgical Scissors
    165 mm (6.5")
    Stainless steel
    Sharp/Blunt
    Each (1UN)
    Each (1UN) - $93.28
    $93.28
    Each (1UN)
       $93.28
     : WLS26975-C
  • Surgical Scissors
    203 mm (8")
    Stainless steel
    Sharp/Blunt
    Each (1UN)
    Each (1UN) - $102.85
    $102.85
    Each (1UN)
       $102.85
     : S26975-D