Support Stands with Rods and Rings
A Convenient Method of Purchasing Both Rings and Supports
- Sets comes with 2, 3, or 4 rings of varied diameters
- Base is stamped steel and the plate underneath is made of steel
- Base weighs 2 lbs.
The support stand will support up to 10 lbs.
Frequently Bought Together