Support Stand with Rod
Supplier: UNITED SCIENTIFIC SUPPLIES
Jump to:
About this item
Weighted With Additional Steel Plate
- Zinc plated rod
These stamped steel bases feature an additional steel plate on the back, which makes them 50% heavier than regular bases. The steel plate also facilitates a sturdier threading of the rod into the base.
2 Options Available Below
Product Details & Documents
Weighted With Additional Steel Plate
- Zinc plated rod
These stamped steel bases feature an additional steel plate on the back, which makes them 50% heavier than regular bases. The steel plate also facilitates a sturdier threading of the rod into the base.