Substage Illuminator
Substage Illuminator
Catalog # 470014-570
Supplier: Swift Instruments
Supplier #: 1101002100012
Jump to:
Product Details & Documents
Inexpensive upgrade for mirror-illuminated microscope.
Add consistent, controlled 15 W tungsten illumination to most mirror microscopes quickly and easily; simply replace the mirror with the substage illuminator, fitting it into the body of the scope. 110 VAC with a 6' 11" two-wire cord and a control switch on the cord.
Size: 41/2L×2W×2"H.
Delivery information: Instructions are included. Replacement bulb sold separately.
Specifications
- Description:Substage Illuminator