Orders LinkContactUsLinkComponent
Student-Grade Polypropylene Beakers
Student-Grade Polypropylene Beakers
Supplier:   SHIVDIAL SUD & SONS
Student-Grade Polypropylene Beakers
Certificates
Student-Grade Polypropylene Beakers
Student-Grade Polypropylene Beakers
6 Options Available Below
Product Details & Documents
These simple, stackable, ribbed beakers with raised and permanent graduations are an excellent value.
  • Polypropylene construction
  • Stackable
  • Simple to use
  • Permanent graduations for easy measurements
  • Autoclavable at 121 °C
Perfect for use with acids, bases, and alcohols, they can also be used short-term with hydrocarbons.
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 50 ml (1.7 oz.)
    Log in for availability
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $7.52
    $7.52
    Log in for availability
    Case de 12 (1UN)
    Case de 12 (1UN) - $1,609.92
    $1,609.92
    Log in for availability
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $8.20
    $8.20
    Each (1UN)
       $7.52
    Case de 12 (1UN)
       $1,609.92
    Paquete de 12 (1UN)
       $8.20
    Supplier Number: CH10155/1
  • 100 ml (3.4 oz.)
    Log in for availability
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $10.77
    $10.77
    Log in for availability
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $11.74
    $11.74
    Log in for availability
    Case de 48 (1UN)
    Case de 48 (1UN) - $516.92
    $516.92
    Each (1UN)
       $10.77
    Paquete de 12 (1UN)
       $11.74
    Case de 48 (1UN)
       $516.92
    Supplier Number: CH10155/3
  • 150 ml (5 oz.)
    Log in for availability
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $132.76
    $132.76
    Log in for availability
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $144.71
    $144.71
    Log in for availability
    Case de 48 (1UN)
    Case de 48 (1UN) - $6,372.58
    $6,372.58
    Each (1UN)
       $132.76
    Paquete de 12 (1UN)
       $144.71
    Case de 48 (1UN)
       $6,372.58
    Supplier Number: F26212-0000_SE
  • 250 ml (8.5 oz.)
    Log in for availability
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $17.20
    $17.20
    Log in for availability
    Case de 40 (1UN)
    Case de 40 (1UN) - $30,613.00
    $30,613.00
    Log in for availability
    Paquete de 40 (1UN)
    Paquete de 40 (1UN) - $18.75
    $18.75
    Each (1UN)
       $17.20
    Case de 40 (1UN)
       $30,613.00
    Paquete de 40 (1UN)
       $18.75
    Supplier Number: CH10155/7
  • 400 ml (14 oz.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,030.00
    $1,030.00
    Log in for availability
    Paquete de 6 (1UN)
    Paquete de 6 (1UN) - $171.09
    $171.09
    Each (1UN)
       $1,030.00
    Paquete de 6 (1UN)
       $171.09
    Supplier Number: F26214-0000
  • 600 ml (20 oz.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $109.65
    $109.65
    Log in for availability
    Paquete de 4 (1UN)
    Paquete de 4 (1UN) - $119.52
    $119.52
    Each (1UN)
       $109.65
    Paquete de 4 (1UN)
       $119.52
    Supplier Number: BP0600