Porcelain Crucibles with Covers
Supplier: AMERICAN SCIENTIFIC LLC SE
Certificates
About this item
These crucibles are great for student use.
- They withstand up to 1000 °C
- Glazed except bottom and rim surfaces
- Acid-resistant and chemically inert
- Great for student use
- Covers included
Except for the bottom and rim surfaces, the crucibles are glazed. They can withstand temperatures up to 1000 °C.
Each cover has a center-ring handle.
Frequently Bought Together