Student Wet Spirometer, 9 L
Disposable Mouthpieces, 2.2 cm
Catalog # 470007-992
Supplier: PHIPPS & BIRD INC.
Número de proveedor #: 7085-301
Jump to:
Product Details & Documents
Increased Volume Size to Accommodate Students with Larger Lung Capacities
Measure Tidal Volume (TV), Inspiratory Capacity (IC), Inspiratory Reserve Volume (IRV), Expiratory Reserve Volume (ERV), and Vital Capacity (VC) with this spirometer that features large, easy-to-read scale graduations. Constructed of durable polyethylene and aluminum, it comes with 100 disposable mouthpieces, a one way plastic valve for sanitary classroom use, instructions, and a lab manual of experiments. Size: 30"H x 18"L x 18"W.
Especificaciones
- Description:Disposable Mouthpieces
- Outer Diameter:2.2 cm