Student Platform Scale
11 lbs. (5000 g)
Catalog # 470106-426
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: KCZ-002
Jump to:
Product Details & Documents
Easy for All Grade Levels to Use.
- Constructed of durable, lightweight plastic
- Large read-out front dial
- Clear protective lens fits over the dial
This desktop scale allows you to measure items up to 5000 grams. Display is in pounds and kilograms.
Specifications
- Weighing Capacity:11 lbs. (5000 g)