Student Plankton Net
Student Plankton Net, 80μm mesh, 20.5", 6", 80 μm
Catalog # 470021-296
Supplier: PECO SALES INC SE
Supplier #: 77919
Jump to:
Product Details & Documents
Perfect for Introductory Plankton Sampling
- Economical
This 20 1/2" Nitex plankton net is attached to a 6" diameter stainless steel ring and bridle. A clear, polycarbonate cod-end assembly with latex tube and pinch clamp and a ten meter calibrated polypropylene line are also included.
Specifications
- Description:Student Plankton Net, 80μm mesh
- Mesh Size:80 μm
- Depth:20.5"
- Width:6"