Student Metric Micrometer
Metric Micrometer, 0 - 25 mm, 0.01 mm
Catalog # 470149-506
Supplier: VWR International
Jump to:
Product Details & Documents
Satin chrome-finished instrument.
- Metric scale
Especificaciones
- Description:Metric Micrometer
- Measurement Range:0 - 25 mm
- Readability:0.01 mm