Ward's® Student Hand Microtome
Ward's® Student Hand Microtome, 1 Pc.
Catalog # 470004-504
Supplier: Avantor
Jump to:
Product Details & Documents
Explore section slide making
- Create your own slides with your own specimens
- Safety features prevent injury
Our unique, easy-to-use microtome features a built-in safety flange to protect fingers from cuts. It includes wax embedding granules, razor knife, five blades, and instructions.
2" dia. platform, 2" length
Documents
Especificaciones
- Description:Ward's® Student Hand Microtome, 1 Pc.