Student Desk Globe
Student Globe, 22.8 cm (9")
Catalog # 470105-304
Supplier: REPLOGLE GLOBES PARTNERS LLC
Número de proveedor #: 50518
Jump to:
Product Details & Documents
This 9" diameter student globe is light enough to pick up and hold.
- Globe detaches from base
- For individual student use
- Mount included
The globe can easily be picked up and held. It is designed for students to use individually at their desks. The globe is painted in seven brilliant colors and furnished with a mount.
Especificaciones
- Description:Student Globe
- Diameter:22.8 cm (9")