Straight Filament Lamp Bulbs
470122-618
- Descripción:Straight Filament Lamp Bulb
- Referencia:470122-618
- Potencia:25 W
- Nº de proveedor:4V461
Single-line filament is ideal for student diffraction experiments. It's also a great way for students to observe filament technology in action, because the slender 15.2 cm lamp lets them clearly and distinctly see the entire process.
Medium screw base and 120 V, suitable for standard sockets.