Stomach, Composite Slide
Stomach, Composite Slide, Section, Hematoxylin and Eosin
470182-856
: Avantor
Compare and contrast sections of this digestive organ
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Three regions: cardiac, fundic, and pyloric.
- Description:Stomach, Composite Slide
- Stain:Hematoxylin and Eosin
- Orientation:Section