Sterile Specimen Container
120 ml (4 oz.), Sterile Specimen Container
Supplier: MCKESSON GENERAL MEDICAL CORP
Número de proveedor #: 491102
Jump to:
Product Details & Documents
Translucent specimen containers with imprinted, screw-on, leak-resistant lid.
- Tamper-evident seal
- Includes patient label
- Not intended for use in pneumatic tubes
- Sterile inner surface only
- Single patient use
- Not made with natural rubber latex
Two graduation scales are marked in 10 cc (mL) and 1/4 oz. increments up to 120 cc. The leak-proof lid has room to write subject information on. Capacity: 4 oz.
Documents
Especificaciones
- Capacity:120 ml (4 oz.)
- Description:Sterile Specimen Container