Sterile Scalpel
Plastic, 165 mm (6.5"), Sterile Scalpel, 6 1/2" No. 22 Blade, #22
Catalog # 470145-792
Supplier: C & A SCIENTIFIC CO INC
Supplier #: 9422
Jump to:
Product Details & Documents
The permanently fitted blades are durable enough to be reused in non-sterile applications, sterilized before the next use, or simply disposed of for convenience.
They come packaged in sealed wrap to protect them from contamination.
Specifications
- Description:Sterile Scalpel, 6 1/2" No. 22 Blade
- Length:165 mm (6.5")
- Material:Plastic
- Blade No.:#22
- Cat. No.:470145-792