ContactUsLinkComponent
Disposable Petri Dishes
  470199-974
undefined
Disposable Petri Dishes
  470199-974
 :  MT2905
  • Ø:
    100 mm
  • Esterilidad:
    Sterile
  • Altura:
    20 mm (0.79")
  • Matriz:
    Polystyrene
  • Referencia:
    470199-974
  • Nº de proveedor:
    1705
Petri Dishes with Covers
 : One package has 20 Petri dishes.