Stentor Slide
Stentor Slide, Whole mount, Schneider's and Fast Green
470177-504
: Avantor
Ciliate protist species
- Identifying features clearly distinguishable
- Rigorous quality control standards
Trumpet-shaped spirotrich. Large macro-nucleus appears in the form of a long chain of beads.
- Description:Stentor Slide
- Stain:Schneider's and Fast Green
- Orientation:Whole mount