Steel Long Form Inclined Plane
Catalog # 470016-236
Supplier: STEINER ENTERPRISES SE
Specifications
- Descripción:Steel Long Form Inclined Plane
- Dimensiones:120 × 13 × 2.5 cm
- Referencia:470016-236
- Nº de proveedor:0813A
Specifications
About this item
This inclined plane is as straight as an arrow, measures a full 4' long, and will last for years. Is built of nickle-plated heavy steel with rounded corners, and can accomondate just about any kind of cart you choose.
- Included: steel long form inclined plane and bucket
- Inclined Plane is 120×13×2.5 cm
- Additional supplies required: car, cord, support stand and rod with clamp and a weight pan or hanger purchased separately
Holes in the supporting flanges fit a 10 mm support rod. Includes instructions and a 40 mm molded styrene pulley on a metal bucket to be mounted in a hole at one end. You will need a cart, cord, support stand and rod with clamp and a weight pan or hanger.