Horseshoe Magnet
Horseshoe Magnet, 3", 2"
Catalog # 470018-704
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: MHS030
Jump to:
Product Details & Documents
Teeny but Strong!
- Keepers helped retain the magnetic strength
- Able to pull 6 pounds
This powerful permanent magnet weighs only 1 oz., but pulls 6 lbs. It comes with a soft iron “keeper” and has a gap of 3/4" at the open end.
Specifications
- Description:Horseshoe Magnet
- Length:3"
- Width:2"