Steel Forceps
Forceps, 130 mm
470017-024
: FRST13
Excellent for Handling Harmful Chemicals or Hot Objects.
- Nickel-plated to prevent rust
- File-cut blunt points
- Points fit squarely together
- Length: 13 cm
Forceps are a valuable tool for handling objects you do not want, or can not touch with your hands.
- Description:Forceps
- Length:130 mm