Static Wand
Static Wand
Catalog # 470003-682
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrate Electrostatic Propulsion and Repulsion
- The set includes a static wand, four static bands, rabbit fur, and teacher’s guide
Rub the rabbit fur over the wand to create a charge, then hold the wand beneath the static band and watch it hover in mid-air, even as you walk around the room.
Especificaciones
- Description:Static Wand