Ward's® Starfish Model
Starfish, 24, 1, 25 cm, 25 cm, 5 cm
Catalog # 470004-388
Supplier: Avantor
Jump to:
Product Details & Documents
Life-Size, Cast From An Actual Specimen
- Highly Durable Rubberized Material
- Common Dissection Organs Highlighted
- Easily Cleaned And Stored
The durable, flexible, freestanding model shows all the intricate details of the starfish’s surface structure as well as its internal anatomy. Three arms are dissected at various levels to show the digestive, reproductive, and water vascular systems; one arm is cross-sectioned to reveal the coelom.
Documents
470004-388 Key
Product Literature
Specifications
- Description:Starfish
- Height:5 cm
- Length:25 cm
- No. of Keyed Items:24
- Width:25 cm
- No. of Parts:1