Spinplus® Magnetic Stirring Bars
Spinplus Magnetic Stir Bar, Plus Sign, 25.4 mm (1"), 14.3 mm (9/16")
Catalog # 470013-464
Supplier: BEL-ART PRODUCTS SE
Número de proveedor #: F37144-0100_SE
Jump to:
Product Details & Documents
Spinplus® magnetic stirring bars add speed and efficiency to mixing operations.
- The “+” shape creates a deep vortex and provides stable, quiet operation
- Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low friction coating; durable and inert
- Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
- Autoclave safe
Especificaciones
- Ø:14.3 mm (9/16")
- Description:Spinplus Magnetic Stir Bar
- Length:25.4 mm (1")
- Shape:Plus Sign
- Cat. No.:470013-464