Standard Magnetic Applicator
Standard Magnetic Applicator, 5.5"
Catalog # 470020-210
Supplier: FORENSICS SOURCE
Número de proveedor #: 1003942
Jump to:
Product Details & Documents
Release Powder Easily, Even Overhead
- Easy Release
- Ergonomical design
A favorite among teachers and students, this ergonomically-designed magnetic applicator has a powerful magnet to hold a large amount of magnetic powder. The spring handle holds the internal magnet in place until you are ready to release the powder allowing for skillful dusting on a variety of surfaces and angles; even overhead! Applicator is also available as part of Ward's Magnetic Fingerprint Kit (36 W 6139).
Especificaciones
- Description:Standard Magnetic Applicator
- Length:5.5"