Orders LinkContactUsLinkComponent
Stake Flags
Stake Flags
Supplier:   PECO SALES INC SE
Stake Flags
Supplier:  PECO SALES INC SE
Certificates
Stake Flags
2 Options Available Below
Product Details & Documents
A versatile marking tool.
  • Aligned with GLOBE protocols
Use these flags attached to steel wire stakes to mark areas for field or watershed studies, or use them for other activities, such as species identification. The flags are solid-colored, colorfast vinyl and flexible. They are also mold and mildew resistant. The flags are 4" x 5", the stakes are 30" high, and they come in packages of 100.
Product Family Options
Información del producto
  • Availability
    Price
  • 470018-560
    Stake Flags, Orange
    Stake Flags, Orange
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $533.53
    $533.53
    Paquete de 100
       $533.53
    Supplier Number: 39549
  • 470018-566
    Stake Flags, Yellow
    Stake Flags, Yellow
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $462.12
    $462.12
    Paquete de 100
       $462.12
    Supplier Number: 39551