ContactUsLinkComponent
GPI Anatomicals® Stages of Osteoarthritis Knee Model
Catalog # 470029-480
Supplier:  GPI ANATOMICALS
Small Business Enterprise
undefined
GPI Anatomicals® Stages of Osteoarthritis Knee Model
Catalog # 470029-480
Supplier:  GPI ANATOMICALS
Supplier Number:  1100
Small Business Enterprise
Specifications
  • Descripción:
    Stages of Osteoarthritis Knee
  • Altura:
    13 cm
  • Longitud:
    8 cm
  • No. of Keyed Items:
    18
  • Referencia:
    470029-480
  • Anchura:
    27 cm
  • Nº de proveedor:
    1100
  • No. of Parts:
    5
Specifications
About this item
Set Of Four Models Depicting Common Disease
Shown in 3/4 scale, this four piece model illustrates the progression of degenerative joint disease. Knee joints representing osteoarthritis, erosion to articular cartilage, and osteophytes or bone spurs at the articular surfaces highlight the stages and severity of the disease.