Square Polystyrene Disposable, Sterile Petri Dishes
Catalog # 470144-494
Supplier: SIMPORT SCIENTIFIC INC.
Specifications
- Descripción:Square Polystyrene Disposable, Sterile Petri Dishes
- Referencia:470144-494
- Nº de proveedor:D210-16
Specifications
About this item
Petri dishes having a raised square grid on the bottom part. The grid consists of 36 small squares; each square is identified to facilitate special type counting or location of specimens. Supplied with top and bottom in sterile shelf packages. Size: 100 x 15mm.