Spirogyra, Vegetative Slide
Spirogyra, Vegetative, Whole mount, Fast Green and Mallory's
Catalog # 470177-424
Supplier: Avantor
Jump to:
Product Details & Documents
Filamentous Green Algae
- Identifying features clearly distinguishable
- Rigorous quality control standards
Note chloroplast in each cell. Usually found in fresh water environments.
Especificaciones
- Description:Spirogyra, Vegetative
- Stain:Fast Green and Mallory's
- Orientation:Whole mount