Orders LinkContactUsLinkComponent
Spirogyra, Vegetative Slide
Spirogyra, Vegetative Slide
Spirogyra, Vegetative Slide, Whole mount, Fast Green and Mallory's
Catalog # 470177-422
Supplier:   Avantor
Spirogyra, Vegetative Slide
Catalog # 470177-422
Supplier:  Avantor
Spirogyra, Vegetative Slide
Spirogyra, Vegetative Slide
Spirogyra, Vegetative Slide, Whole mount, Fast Green and Mallory's
Price
$116.28
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Investigate spirogyra structures
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Cells with several chloroplasts; also showing nucleus, pyrenoids, and other cytoplasmic detail.
Especificaciones
  • Description:
    Spirogyra, Vegetative Slide
  • Stain:
    Fast Green and Mallory's
  • Orientation:
    Whole mount