Spinal Cord Slide
Spinal Cord, Cross section, Silver impreg.
Catalog # 470182-706
Supplier: Avantor
Jump to:
Product Details & Documents
Central Nervous System
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Staining method clearly defines gray and white matter as well as large motor nerve cells in ventral horn.
Especificaciones
- Description:Spinal Cord
- Stain:Silver impreg.
- Orientation:Cross section