Spinal Cord, Human Slide
Spinal Cord, Human, Cross section, Cresyl Violet
Catalog # 470182-988
Supplier: Avantor
Jump to:
Product Details & Documents
Central Nervous System
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Shows Nissl bodies. The spinal cord is protected within the spine and connects the brain to other parts of the body.
Especificaciones
- Description:Spinal Cord, Human
- Stain:Cresyl Violet
- Orientation:Cross section