Spinal Cord Slide
Spinal Cord, Cross section & longitudinal section, Cresyl Violet
Catalog # 470182-712
Supplier: Avantor
Jump to:
Product Details & Documents
Central Nervous System
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Fiber bundles and single fiber show up clearly. The nerve cells are well stained and differentiated.
Especificaciones
- Description:Spinal Cord
- Stain:Cresyl Violet
- Orientation:Cross section & longitudinal section