Spinal Cord (Human), Thoracic Level Slide
Spinal Cord (Human), Thoracic Level, Cross section, Hematoxylin and Eosin
Catalog # 470176-968
Supplier: Avantor
Jump to:
Product Details & Documents
Central Nervous System
- Identifying features clearly distinguishable
- Rigorous quality control standards
Thoracic level cross section showing white and grey matter.
Specifications
- Description:Spinal Cord (Human), Thoracic Level
- Stain:Hematoxylin and Eosin
- Orientation:Cross section