Orders LinkContactUsLinkComponent
Spherical Mirrors
Supplier:   VWR International
Spherical Mirrors
Supplier:  VWR International
Certificates
Spherical Mirrors
Spherical Mirrors
16 Options Available Below
Product Details & Documents
These wide range of mirrors are perfect for investigating real and virtual images.
  • Variety of Sizes and Focal Lengths
  • Circular
  • Polished Glass Construction
  • Durable
  • Back-Surface Silvered
The circular mirrors are available in diameters of 38 mm, 50 mm, and 75 mm.
They're back-surface silvered, made of polished glass, and painted for durability.
Product Family Options
Concave
Product Information
  • Focal Length
    Diameter
    Availability
    Price
  • 470014-718
    Concave mirror
    Concave mirror
    50 mm
    38 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $41.57
    $41.57
    Each (1UN)
       $41.57
  • 470111-298
    Concave mirror
    Concave mirror
    100 mm
    38 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $60.21
    $60.21
    Each (1UN)
       $60.21
  • 470111-300
    Concave mirror
    Concave mirror
    150 mm
    38 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $55.44
    $55.44
    Each (1UN)
       $55.44
  • 470111-302
    Concave mirror
    Concave mirror
    250 mm
    38 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $55.44
    $55.44
    Each (1UN)
       $55.44
  • 470013-310
    Concave mirror
    Concave mirror
    300 mm
    50 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $0.75
    $0.75
    Clearance
    Each (1UN)
       $0.75
  • 470013-308
    Concave mirror
    Concave mirror
    200 mm
    50 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $0.90
    $0.90
    Clearance
    Each (1UN)
       $0.90
  • 470013-306
    Concave mirror
    Concave mirror
    150 mm
    50 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $29.32
    $29.32
    Each (1UN)
       $29.32
  • 470013-304
    Concave mirror
    Concave mirror
    100 mm
    50 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $29.32
    $29.32
    Each (1UN)
       $29.32
  • 470016-996
    Concave mirror
    Concave mirror
    70 mm
    75 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $85.28
    $85.28
    Each (1UN)
       $85.28
Convex
Product Information
  • Focal Length
    Diameter
    Availability
    Price
  • 470005-646
    Convex mirror
    Convex mirror
    –50 mm
    38 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $46.91
    $46.91
    Each (1UN)
       $46.91
  • 470111-304
    Convex mirror
    Convex mirror
    –100 mm
    38 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $60.21
    $60.21
    Each (1UN)
       $60.21
  • 470111-306
    Convex mirror
    Convex mirror
    –150 mm
    38 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $55.44
    $55.44
    Each (1UN)
       $55.44
  • 470111-308
    Convex mirror
    Convex mirror
    –250 mm
    38 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $55.44
    $55.44
    Each (1UN)
       $55.44
  • 470013-312
    Convex mirror
    Convex mirror
    –100 mm
    50 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $29.32
    $29.32
    Each (1UN)
       $29.32
  • 470013-314
    Convex mirror
    Convex mirror
    –150 mm
    50 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $29.32
    $29.32
    Each (1UN)
       $29.32
  • 470013-316
    Convex mirror
    Convex mirror
    –200 mm
    50 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $26.66
    $26.66
    Each (1UN)
       $26.66