Orders LinkContactUsLinkComponent
Spectrum Tubes
Spectrum Tubes
Spectrum Tubes
Spectrum Tubes
Spectrum Tubes
Spectrum Tubes demonstrate the wave properties of different gases.
  • Ideal for Wavelength Determination
  • For Introducing Students to Better-Known Gases
  • Glass Tube Length: 10"
  • Capillary Portion Length: 4"
  • Tubes Work in a Variety of Spectroscopic Applications
Ideal for wavelength determination and introducing students to the spectra of better-known gases, these glass tubes are 10" long, and the capillary portion is about 4" long. The spectrum tube assemblies can be used in a variety of spectroscopic applications.
When they are observed through a spectroscope, the emission lines corresponding to the energy transition levels of the gaseous atoms or molecules appear as distinct components. The Spectrum Tube Power Supply is required but not included.
Ordering information: The mercury spectrum tube contains mercury and may not be available for sale in your state. Contact customer service for details.
Product Family Options
  • Description
    Length
  • Air
    25.4 cm
    Each (1UN)
    Each (1UN) - $399.75
    $399.75
    Each (1UN)
       $399.75
  • Argon
    25.4 cm
    Each (1UN)
    Each (1UN) - $401.34
    $401.34
    Each (1UN)
       $401.34
  • Carbon Dioxide
    25.4 cm
    Each (1UN)
    Each (1UN) - $399.75
    $399.75
    Each (1UN)
       $399.75
  • Chlorine
    25.4 cm
  • Helium
    25.4 cm
    Each (1UN)
    Each (1UN) - $399.75
    $399.75
    Each (1UN)
       $399.75
  • Hydrogen
    25.4 cm
    Each (1UN)
    Each (1UN) - $399.75
    $399.75
    Each (1UN)
       $399.75
  • 50-50 mix H and He
    25.4 cm
    Each (1UN)
    Each (1UN) - $399.75
    $399.75
    Each (1UN)
       $399.75
  • Iodine Vapor
    25.4 cm
    Each (1UN)
    Each (1UN) - $399.75
    $399.75
    Each (1UN)
       $399.75
  • Krypton
    25.4 cm
    Each (1UN)
    Each (1UN) - $399.75
    $399.75
    Each (1UN)
       $399.75
  • Neon
    25.4 cm
    Each (1UN)
    Each (1UN) - $399.75
    $399.75
    Each (1UN)
       $399.75
  • Nitrogen
    25.4 cm
    Each (1UN)
    Each (1UN) - $399.75
    $399.75
    Each (1UN)
       $399.75
  • Oxygen
    25.4 cm
    Each (1UN)
    Each (1UN) - $399.75
    $399.75
    Each (1UN)
       $399.75
  • Deuterium
    25.4 cm
    Each (1UN)
    Each (1UN) - $746.21
    $746.21
    Each (1UN)
       $746.21
  • Water Vapor
    25.4 cm
    Each (1UN)
    Each (1UN) - $399.75
    $399.75
    Each (1UN)
       $399.75
  • Xenon
    25.4 cm
    Each (1UN)
    Each (1UN) - $399.75
    $399.75
    Each (1UN)
       $399.75
  • Sulphur
    Each (1UN)
    Each (1UN) - $770.19
    $770.19
    Each (1UN)
       $770.19