CENCO® Electrodeless Spectrum Tube Power Supply
Supplier: STEINER ENTERPRISES SE
Certificates
About this item
This safe spectrum power supply features encased spectrum tubes.
- No Exposure to Voltage Source
- No Electrodes in Bulbs
- All Spectrum Tubes Are Encased Inside the Power Supply
- For Use with Spectroscopes or Spectrometers
- Bulbs Have 150 Hours of Burn Time
This innovative device far surpasses the safety, capability, and convenience of any other spectrum power supply available.
Most current power supplies expose students to 5000V. This device’s new, advanced design and features eliminate student’s contact to the voltage source. This safety feature allows you to focus attention on your students' learning experience instead of worrying about their safety.
Frequently Bought Together