Specimen Tongs
Tongs, Flask & Test Tube, 20 cm long
Catalog # 470005-898
Supplier: DUNRITE INSTRUMENTS SE
Número de proveedor #: 6343500
Jump to:
Product Details & Documents
Ideal for handling preserved specimens, the wide clamping jaws are flattened and have serrations to prevent slipping.
The jaw spread is 2 inches. The tongs are constructed of nickel-plated steel and have an overall length of 8 inches.
Especificaciones
- Description:Tongs, Flask & Test Tube, 20 cm long