Orders LinkContactUsLinkComponent
Specific Gravity Hydrometer
Specific Gravity Hydrometer
Supplier:   G&W INSTRUMENTS
Specific Gravity Hydrometer
Supplier:  G&W INSTRUMENTS
Certificates
Specific Gravity Hydrometer
Specific Gravity Hydrometer
2 Options Available Below
Product Details & Documents
Use these hydrometers with all processes where specific gravity or density of liquids is an important control.
The design of this hydrometer ensures easy cleaning and a low likelihood of breakage. The clear markings allow accurate readings. Accuracy: ±0.001. Size: 12"L.
Product Family Options
Información del producto
  • Availability
    Price
  • 470005-562
    Specific Gravity Hydrometer, 0.70–1.00, 0.005 divisions
    Specific Gravity Hydrometer, 0.70–1.00, 0.005 divisions
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $8.90
    $8.90
    Liquidación
    Each (1UN)
       $8.90
    Supplier Number: A-1177
  • 470019-566
    Specific Gravity Hydrometer, 1.00–1.20, 0.002 divisions
    Specific Gravity Hydrometer, 1.00–1.20, 0.002 divisions
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $313.40
    $313.40
    Each (1UN)
       $313.40
    Supplier Number: A-1101