Somso® Teeth Model Set
Teeth Set, 8, 1
Catalog # 470029-924
Supplier: MARCUS SOMMER
Número de proveedor #: ES 11
Jump to:
Product Details & Documents
Large, Up-Close View Of Each Type Of Tooth
- Each Model Is Represented On Its Own Base
- Lower Molar Includes Caries
- 5 Year Manufacturers Warranty
Five different models illustrate every major tooth type. The lower incisor features a removable half of the crown, the lower canine separates into two longitudinally, the lower molar with one root is one-piece, the lower molar with two roots separates into three parts showing dental caries in various stages, and the first upper molar with three roots separates into three parts. Each model is magnified to 8X and is mounted on its own stand with a base.
Documents
Somso Production Film
Videos
Especificaciones
- Description:Teeth Set
- No. of Keyed Items:8
- No. of Parts:1