Solar Motion Model
Catálogo # 470004-710
Proveedor: THE SCIENCE SOURCE SE
Especificaciones
- Descripción:Solar Motion Model
- Referencia:470004-710
- Nº de proveedor:10240
Especificaciones
Acerca de este artículo
Show the Basis for the Seasons and Solar Behavior
- Detailed solar model Explains Earth’s rotation and orbit Hands-on learning experience 12" diameter
Visualize the Sun’s Journey with the Solar Motion Model. This hands-on model offers students a clear, interactive way to explore how Earth’s rotation and orbit shape the cycle of day and night. By simulating the Sun’s path across the sky, learners can observe how solar motion varies by location and season, deepening their understanding of Earth’s axial tilt and the changing seasons. Designed for both educational value and visual appeal, the model features a sturdy wood disk and a stand, making it a functional and attractive addition to any classroom, desktop, or library display.