Solar Electricity Kit
Kit, Solar Electricity, 1
Catalog # 470004-940
Supplier: SOLAR MADE SE
Número de proveedor #: SK-50
Jump to:
Product Details & Documents
With this Solar Cell Kit, you can harness the power of the sun.
- Experiment with natural and artificial light
- Use solar power to run several devices
- Includes an instruction manual
This Solar Energy Kit consists of an encapsulated solar cell array, a miniature motor, a wind propeller, an optical illusion disc and an instruction manual.
Students can create a solar desk fan and a "virtual spiral" device, study the impact of artificial light versus sunlight and conduct numerous other experiments using the solar cell.
Especificaciones
- Description:Kit, Solar Electricity
- No. of Lab Groups:1