Solar Cell
Solar Cells
Catalog # 470002-992
Supplier: VWR International
Jump to:
Product Details & Documents
Show How Direct/Indirect Sunlight Provides Power
A photo-voltaic cell, this solar cell uses illumination to produce a voltage able to drive current through a load. The cell used in this device is incredibly sensitive and capable of delivering 100mW into a 10 load when a 36W filament bulb is held approximately 30 mm from the cell.
Especificaciones
- Description:Solar Cells