Soil pH Meter
Rapitest Soil pH Meter
Catalog # 470104-844
Supplier: LUSTER LEAF PRODUCTS INC SE
Número de proveedor #: 1840
Jump to:
Product Details & Documents
Simple read out.
- Easy use
- Indoor or outdoor
- Aligned with GLOBE protocols
Provide proper conditions for plant growth with this easy-to-use meter, which instantly measures soil pH from 3.5 to 9.0. The supplied reference chart lists preferred pH for more than 400 vegetables, herbs, fruits, flowers, trees, shrubs and house plants, and offers advice for keeping plants healthy.
Includes: teacher’s guide with instructions, background information and recommendations for correcting pH.
Especificaciones
- Description:Rapitest Soil pH Meter
- Cat. No.:470104-844
- pH range:3.5 - 9.0