Soft Iron Rod
Proveedor: STEINER ENTERPRISES SE
Certificados
Acerca de este artículo
Soft iron is a classic example of a readily magnetized material.
- For Magnetic Demonstrations
- Low-Remanence
- Low-Coercivity
- Easy-To-Magnetize
- Ten Rods per Package
Since it has low remanence and coercivity, it is also readily demagnetized.
This soft iron rod is ideal for magnetic demonstrations.
Embalaje: This item is available in packages of 10 pieces.
Frequently Bought Together