Smooth Muscle, Thin Section Slide
Smooth Muscle, Thin Section Slide, Thin Section, Hematoxylin and Eosin
Catalog # 470183-664
Supplier: Avantor
Jump to:
Product Details & Documents
Extremely Thin Plastic Embedded Tissue For Greater Viewing Clarity
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal intestine. Elongated, spindle-shaped cells containing a single nucleus. It illustrates typical structures.
Especificaciones
- Description:Smooth Muscle, Thin Section Slide
- Stain:Hematoxylin and Eosin
- Orientation:Thin Section