Smilax, Root Slide
Smilax, Root, Cross section, Quadruple
Catalog # 470181-278
Supplier: Avantor
Jump to:
Product Details & Documents
Greenbrier
- Identifying features clearly distinguishable
- Rigorous quality control standards
Greenbrier. Shows radial arrangement of xylem with heavy endodermis.
Especificaciones
- Description:Smilax, Root
- Stain:Quadruple
- Orientation:Cross section